Merck
A missense mutation W797R in the myophosphorylase gene in a Spanish patient with McArdle's disease.
Muscle & nerve (1999-12-11)
J C Rubio, M A Martín, Y Campos, R Auciello, A Cabello, J Arenas
ABSTRAKT
We identified a novel missense mutation in the myophosphorylase gene (PYGM) in a Spanish patient with McArdle's disease. This homozygous T-to-C transition results in the replacement of a highly conserved tryptophan at amino acid position (aa) 797 with an arginine in the C-terminal domain of the PYGM protein. The lack of enzyme activity in the proband's muscle is consistent with a crucial role of the aa 797 in the normal function of the PYGM protein. Our data further expand the genetic heterogeneity in patients with McArdle's disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosphorylase a from rabbit muscle, lyophilized powder, 20-30 units/mg protein
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization