Merck
Human epidermis is a novel site of phospholipase B expression.
Biochemical and biophysical research communications (2002-08-02)
Eric Maury, Marie Claude Prévost, Michel Nauze, Daniel Redoulès, Roger Tarroux, Marie Charvéron, Jean Pierre Salles, Bertrand Perret, Hugues Chap, Ama Gassama-Diagne
ABSTRAKT
Phospholipase B (PLB) is an enzyme that displays both phospholipase A(2) and lysophospholipase activities. Analysis of human epidermis homogenates indicated the presence of a 97 kDa PLB protein, as well as a phospholipase A(2) activity, both being enriched in the soluble fraction. Immunolabelling and in situ hybridization experiments showed that this enzyme is expressed in the different layers of epidermis with an accumulation at the dermo-epidermis junction. RT-PCR data indicated that PLB is specifically expressed in natural and reconstructed epidermis. By 3'-RACE-PCR and screening of human genome databases, we obtained a 3600 bp cDNA coding for human PLB highly homologous to already described intestinal brush border PLBs. These data led us to conclude that the soluble PLB corresponds to a proteolytic cleavage of the membrane anchored protein. Altogether, our results provide the first characterization of human PLB which should play an important role in epidermal barrier function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein