Merck
Effects of human parvovirus B19 VP1 unique region protein on macrophage responses.
Journal of biomedical science (2009-03-11)
Bor-Show Tzang, Chun-Ching Chiu, Chun-Chou Tsai, Yi-Ju Lee, I-Jung Lu, Jing-Yu Shi, Tsai-Ching Hsu
ABSTRAKT
Activity of secreted phospholipase A (sPLA2) has been implicated in a wide range of cellular responses. However, little is known about the function of human parvovirus B19-VP1 unique region (VP1u) with sPLA2 activity on macrophage. To investigate the roles of B19-VP1u in response to macrophage, phospholipase A2 activity, cell migration assay, phagocytosis activity, metalloproteinase assay, RT-PCR and immunoblotting were performed. In the present study, we report that migration, phagocytosis, IL-6, IL-1beta mRNA, and MMP9 activity are significantly increased in RAW264.7 cells by B19-VP1u protein with sPLA2 activity, but not by B19-VP1uD175A protein that is mutated and lacks sPLA2 activity. Additionally, significant increases of phosphorylated ERK1/2 and JNK proteins were detected in macrophages that were treated with B19-VP1u protein, but not when they were treated with B19-VP1uD175A protein. Taken together, our experimental results suggest that B19-VP1u with sPLA2 activity affects production of IL-6, IL-1beta mRNA, and MMP9 activity, possibly through the involvement of ERK1/2 and JNK signaling pathways. These findings could provide clues in understanding the role of B19-VP1u and its sPLA2 enzymatic activity in B19 infection and B19-related diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Giemsa stain, technical grade, used as a blood stain
Sigma-Aldrich
Giemsa stain, certified by the BSC