Merck
Transcriptional mutagenesis induced by 8-oxoguanine in mammalian cells.
PLoS genetics (2009-07-25)
Damien Brégeon, Paul-Antoine Peignon, Alain Sarasin
ABSTRAKT
Most of the somatic cells of adult metazoans, including mammals, do not undergo continuous cycles of replication. Instead, they are quiescent and devote most of their metabolic activity to gene expression. The mutagenic consequences of exposure to DNA-damaging agents are well documented, but less is known about the impact of DNA lesions on transcription. To investigate this impact, we developed a luciferase-based expression system. This system consists of two types of construct composed of a DNA template containing an 8-oxoguanine, paired either with a thymine or a cytosine, placed at defined positions along the transcribed strand of the reporter gene. Analyses of luciferase gene expression from the two types of construct showed that efficient but error-prone transcriptional bypass of 8-oxoguanine occurred in vivo, and that this lesion was not repaired by the transcription-coupled repair machinery in mammalian cells. The analysis of luciferase activity expressed from 8OG:T-containing constructs indicated that the magnitude of erroneous transcription events involving 8-oxoguanine depended on the sequence contexts surrounding the lesion. Additionally, sequencing of the transcript population expressed from these constructs showed that RNA polymerase II mostly inserted an adenine opposite to 8-oxoguanine. Analysis of luciferase expression from 8OG:C-containing constructs showed that the generated aberrant mRNAs led to the production of mutant proteins with the potential to induce a long-term phenotypical change. These findings reveal that erroneous transcription over DNA lesions may induce phenotypical changes with the potential to alter the fate of non-replicating cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TRI Reagent®
Sigma-Aldrich
TRI Reagent®, BD, For processing whole blood, plasma, or serum.
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
TRI Reagent®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.