Merck
CD83 increases MHC II and CD86 on dendritic cells by opposing IL-10-driven MARCH1-mediated ubiquitination and degradation.
The Journal of experimental medicine (2011-01-12)
Lina E Tze, Keisuke Horikawa, Heather Domaschenz, Debbie R Howard, Carla M Roots, Robert J Rigby, David A Way, Mari Ohmura-Hoshino, Satoshi Ishido, Christopher E Andoniou, Mariapia A Degli-Esposti, Christopher C Goodnow
ABSTRAKT
Effective vaccine adjuvants must induce expression of major histocompatibility (MHC) class II proteins and the costimulatory molecule CD86 on dendritic cells (DCs). However, some adjuvants elicit production of cytokines resulting in adverse inflammatory consequences. Development of agents that selectively increase MHC class II and CD86 expression without triggering unwanted cytokine production requires a better understanding of the molecular mechanisms influencing the production and degradation of MHC class II and CD86 in DCs. Here, we investigate how CD83, an immunoglobulin protein expressed on the surface of mature DCs, promotes MHC class II and CD86 expression. Using mice with an N-ethyl-N-nitrosourea-induced mutation eliminating the transmembrane (TM) region of CD83, we found that the TM domain of CD83 enhances MHC class II and CD86 expression by blocking MHC class II association with the ubiquitin ligase MARCH1. The TM region of CD83 blocks interleukin 10-driven, MARCH1-dependent ubiquitination and degradation of MHC class II and CD86 in DCs. Exploiting this posttranslational pathway for boosting MHC class II and CD86 expression on DCs may provide an opportunity to enhance the immunogenicity of vaccines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, FITC conjugate