Merck
Crystal structure of a calcium-phospholipid binding domain from cytosolic phospholipase A2.
The Journal of biological chemistry (1998-01-27)
O Perisic, S Fong, D E Lynch, M Bycroft, R L Williams
ABSTRAKT
Cytosolic phospholipase A2 (cPLA2) is a calcium-sensitive 85-kDa enzyme that hydrolyzes arachidonic acid-containing membrane phospholipids to initiate the biosynthesis of eicosanoids and platelet-activating factor, potent inflammatory mediators. The calcium-dependent activation of the enzyme is mediated by an N-terminal C2 domain, which is responsible for calcium-dependent translocation of the enzyme to membranes and that enables the intact enzyme to hydrolyze membrane-resident substrates. The 2.4-A x-ray crystal structure of this C2 domain was solved by multiple isomorphous replacement and reveals a beta-sandwich with the same topology as the C2 domain from phosphoinositide-specific phospholipase C delta 1. Two clusters of exposed hydrophobic residues surround two adjacent calcium binding sites. This region, along with an adjoining strip of basic residues, appear to constitute the membrane binding motif. The structure provides a striking insight into the relative importance of hydrophobic and electrostatic components of membrane binding for cPLA2. Although hydrophobic interactions predominate for cPLA2, for other C2 domains such as in "conventional" protein kinase C and synaptotagmins, electrostatic forces prevail.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein