Merck
N6-bicycloalkyladenosines with unusually high potency and selectivity for the adenosine A1 receptor.
Journal of medicinal chemistry (1989-01-01)
B K Trivedi, A J Bridges, W C Patt, S R Priebe, R F Bruns