Merck
  • [Pulmonary diagnosis with model aerosols. II: Aerosol bolus dispersion--a marker for convection gas transport. Basic principles and initial clinical results].
[Pulmonary diagnosis with model aerosols. II: Aerosol bolus dispersion--a marker for convection gas transport. Basic principles and initial clinical results].
Pneumologie (Stuttgart, Germany) (1999-03-26)
M Kohlhäufl, P Brand, G Scheuch, H Schulz, K Häussinger, J Heyder
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, technical grade, 90%
Supelco
Bis(2-ethylhexyl) sebacate, Selectophore, ≥97.0%
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, ≥97.0% (GC)