Merck
Chemiluminescence of ABEI-labelled IgG triggered by the N-chloramine-H(2)O(2)-p-iodophenol system.
Luminescence : the journal of biological and chemical luminescence (1999-07-29)
S Olszowski, E Olszowska, T Stelmaszyńska, A Krawczyk
ABSTRAKT
Light is emitted in systems containing N-chloramines or hypochlorite, H(2)O(2) and N-(4-aminobutyl)-N-ethylisoluminol (ABEI). The emission is enhanced by 4-iodophenol (PIP) in alkaline solution (1 mol/L NaOH), while at lower pH range (9-11) PIP is not only inactive but also its presence reduces chemiluminescence (CL) of the monochloramine-H(2)O(2)-ABEI system to the background. Two procedures for ABEI-labelled IgG assays were developed, with PIP in 1 mol/L NaOH and without PIP at pH 11, and the standard curves of free ABEI in these conditions were examined. We suggest also that the oxidative deamination of taurine chloramine leads to the formation of the various carbonyl derivatives and their formation is accelerated in the presence of H(2)O(2), especially in less alkaline solutions (pH 11). Moreover, the formation of enol forms of aldehydes in assay buffers was observed. The yield of the phenoxy radical mediators of ABEI oxidation and the pH-dependent H(2)O(2):HO(2) ratio seems to be decisive for the overall CL in the system examined. The main advantage of this method is that CL does not need precise timing of measurements and assays can be performed over a long period of time (hours) using a plate luminometer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%
Sigma-Aldrich
4-Iodophenol, 99%