Merck
Plasmodium berghei: induction of aminopeptidase in malaria-resistant strain of Anopheles gambiae.
Experimental parasitology (1999-09-30)
A Rosenfeld, J P Vanderberg
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Leucine-p-nitroanilide, leucine aminopeptidase substrate