- Urticaria and hidden salicylates.
Urticaria and hidden salicylates.
Pediatric dermatology (2000-01-13)
A L Wright, A Minford
PMID10632946
ABSTRAKT
A 23-month-old girl had a chronic urticaria resistant to antihistamine therapy. She was found to be using large quantities of a topical salicylate preparation which was being regularly applied to her pacifier. Once this habit was stopped, her urticaria cleared and has not recurred.