Merck
Propachlor removal by Pseudomonas strain GCH1 in an immobilized-cell system.
Applied and environmental microbiology (2000-03-04)
M Martín, G Mengs, E Plaza, C Garbi, M Sánchez, A Gibello, F Gutierrez, E Ferrer
ABSTRAKT
A bacterial strain capable of growing on propachlor (2-chloro-N-isopropylacetanilide) was isolated from soil by using enrichment and isolation techniques. The strain isolated, designated GCH1, was classified as a member of the genus Pseudomonas. Washed-cell suspensions of strain GCH1 accumulated N-isopropylacetanilide, acetanilide, acetamide, and catechol. Pseudomonas strain GCH1 grew on propachlor with a generation time of 4.2 h and a rate of substrate utilization of 1.75 +/- 0.15 micromol h(-1). Gene expression did not require induction but was subject to catabolite expression. Acetanilide was a growth substrate with a yield of 0.56 +/- 0.02 mg of protein micromol(-1). GCH1 strain cells were immobilized by adsorption onto a ceramic support and were used as biocatalysts in an immobilized cell system. Propachlor elimination reached 98%, with a retention time of 3 h and an initial organic load of 0.5 mM propachlor. The viability of immobilized cells increased 34-fold after 120 days of bioreactor operation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Propachlor, PESTANAL®, analytical standard