Merck
Immunohistochemical myofiber typing and high-resolution myofibrillar lesion detection in LR white embedded muscle.
Microscopy research and technique (2000-06-22)
J L Thompson, K Vijayan, D A Riley
ABSTRAKT
We have developed a method of fixing, embedding, sectioning, and staining that allows high-resolution detection of myofibrillar structure and myosin immunocytochemical muscle fiber typing in serial semithin sections of LR White plastic embedded muscle at the light microscopic level. Traditional approaches, such as cryostat sections, permit fiber typing, but small myofibrillar lesions (1-3 sarcomeres) are difficult to detect because of section thickness. Semithin sections of hydrophobic resins do not stain well either histochemically or immunocytochemically. Electron microscopy can resolve lesions and discriminate fiber types based on morphology, but the sampling area is small. Our goal was to develop a rapid method for defining both fiber type and high-resolution primary myofibrillar lesion damage. Mild fixation (1-4% paraformaldehyde, 0. 05-0.1% glutaraldehyde) and embedment in a hydrophilic resin (LR White) were used. Myofibrillar structure was extremely well preserved at the light microscopic (LM) level, and lesions could be readily resolved in Toluidine blue stained 500-nm sections. Fiber type was defined by LM immunomyosin staining of serial plastic semithin sections, which demonstrated reciprocal staining patterns for "fast (Sigma M4276) and "total" (skeletal muscle) myosins (Sigma M7523).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
LR white acrylic resins, Medium