- Syntheses and binding affinities of 6-nitroquipazine analogues for serotonin transporter. Part 1.
Syntheses and binding affinities of 6-nitroquipazine analogues for serotonin transporter. Part 1.
Bioorganic & medicinal chemistry letters (2000-07-29)
B S Lee, S Chu, B C Lee, D Y Chi, Y S Choe, K J Jeong, C Jin
PMID10915050
ABSTRAKT
6-Nitroquipazine has been known as one of the most potent and selective inhibitors of serotonin transporter in vitro and in vivo. Nine derivatives of 6-nitroquipazine were synthesized and tested for their potential abilities to displace [3H]citalopram binding to the rat cortical membranes.