Merck
Spill-induced sensitization to isophoronediamine.
Contact dermatitis (2000-08-17)
D Kelterer, A Bauer, P Elsner
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans, ≥99%