Merck
  • DNA-protein crosslinks induced by nickel compounds in isolated rat lymphocytes: role of reactive oxygen species and specific amino acids.
DNA-protein crosslinks induced by nickel compounds in isolated rat lymphocytes: role of reactive oxygen species and specific amino acids.
Toxicology and applied pharmacology (2001-02-13)
S K Chakrabarti, C Bai, K S Subramanian
ABSTRAKT
Isolated rat lymphocytes in salts-glucose medium (pH 7.2) were incubated with nickel chloride, nickel acetate, nickel sulfate, and a soluble form of nickel subsulfide (0-2 mM) at 37 degrees C for 2 h. The soluble form of nickel subsulfide induced a significant increase in DNA-protein crosslinks (DPXLs) (111%) beginning at 0.5 mM and a maximum increase of 700% from that of the control value was reached at a 2 mM concentration, whereas nickel sulfate produced only a 65% increase of such crosslinks at the 2 mM concentration only. No significant reduction in viability of rat lymphocytes (as measured by trypan blue exclusion) due to these nickel compounds was observed at any concentration used. Time-course studies of DPXLs and cellular viability due to 2 mM nickel subsulfide indicate that DPXL formation may not be due in part to cellular necrosis. Coincubation of nickel subsulfide (2 mM) with l-histidine (16 mM), l-cysteine (4 or 8 mM), or l-aspartic acid (24 mM) significantly reduced the DPXLs induced by 2 mM nickel subsulfide. But Mg(2+) even at 24 mM failed to antagonize nickel subsulfide-induced increase in DPXLs. High concentrations of these amino acids significantly decreased the accumulation of Ni(2+) from nickel subsulfide in lymphocytes, suggesting that such reduction of cellular uptake of Ni(2+) by these amino acids is partly responsible for the potent protective effects of these amino acids against such genotoxicity of nickel subsulfide. In vitro exposure of lymphocytes to nickel subsulfide (0-2 mM) increased the formation of reactive oxygen species (ROS) in a concentration-dependent manner. Furthermore, coincubation of 2 mM nickel subsulfide with catalase, dimethylthiourea, mannitol, or vitamin C at 37 degrees C for 2 h resulted in a significant decrease of nickel subsulfide-induced formation of DPXLs, suggesting that nickel subsulfide-induced DPXLs formation in isolated rat lymphocytes is caused by the formation of ROS. The amino acid treatment also abrogated Ni(3)S(2)-induced generation of ROS. Deferoxamine (a highly specific iron chelator) treatment prevented nickel subsulfide-induced DNA-protein crosslink formation, suggesting that Ni(2+)-induced DPXL formation in rat lymphocytes is caused by the induction of Fenton/Haber-Weiss reaction, generating hydroxyl radicals. The potent protective effects of these specific amino acids against nickel subsulfide-induced DPXL formation in isolated rat lymphocytes may be due in part to impaired cellular uptake of Ni(2+), inhibition of the binding of Ni(2+) to deproteinized DNA, and a reduction in reactive oxygen species.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nickel sulfide, 99.7% trace metals basis, −200 mesh