Merck
High-performance liquid chromatographic determination of methyl anthranilate, hydroxymethylfurfural and related compounds in honey.
Journal of chromatography. A (2001-06-14)
M J Nozal, J L Bernal, L Toribio, J J Jiménez, M T Martín
ABSTRAKT
A high-performance liquid chromatographic method for determining 5-hydroxymethyl-2-furaldehyde (hydroxymethylfurfural), 2-furaldehyde (furfural), furan-2-carboxylic acid (2-furoic acid), furan-3-carboxylic acid (3-furoic acid), furan-3-carboxaldehyde (3-furaldehyde) and 2-aminobenzoic acid methyl ester (methyl anthranilate) in honey and honeydew samples is described. To prevent matrix interference and to isolate the compounds, a clean-up step which implies a solid-phase extraction on polymeric cartridges and an elution with 0.5 ml methanol is recommended. The compounds are separated on a reversed-phase column with a gradient of (A) 1% aqueous acetic acid-acetonitrile (97:3, v/v) and (B) acetonitrile-water (50:50, v/v), with UV detection at 250 nm. The method is applied to the analysis of samples from different botanical origin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyl anthranilate, ≥98%, FCC, FG
Sigma-Aldrich
Methyl 2-aminobenzoate, ReagentPlus®, ≥99%
Sigma-Aldrich
3-Furoic acid, 98%