Merck
  • Phytoestrogens from the roots of Polygonum cuspidatum (Polygonaceae): structure-requirement of hydroxyanthraquinones for estrogenic activity.
Phytoestrogens from the roots of Polygonum cuspidatum (Polygonaceae): structure-requirement of hydroxyanthraquinones for estrogenic activity.
Bioorganic & medicinal chemistry letters (2001-07-19)
H Matsuda, H Shimoda, T Morikawa, M Yoshikawa
ABSTRAKT
The methanolic extract from the roots of Polygonum (P.) cuspidatum was found to enhance cell proliferation at 30 or 100 microg/mL in MCF-7, an estrogen-sensitive cell line. By bioassay-guided separation from P. cuspidatum with the most potent activity, emodin and emodin 8-O-beta-D-glucopyranoside were isolated as active principles. The methanolic extracts from Polygonum, Cassia, Aloe, and Rheum species, which were known to contain anthraquinones, also showed the MCF-7 proliferation. As a result of the evaluation of various anthraquinones from plant sources and synthetic anthraquinones, aloe-emodin, chrysophanol, chrysophanol 8-O-beta-D-glucopyranoside, and 1,8-dihydroxyanthraquinone showed weak activity. On the other hand, alizalin and 2,6-dihydroxyanthraquinone as well as emodin having the 2- and/or 6-hydroxyl groups showed potent activity. These results show that the unchelated hydroxyl group is essential for strong activity. Emodin and 2,6-dihydroxyanthraquinone also inhibited 17beta-estradiol binding to human estrogen receptors (ERs) with K(i) values of 0.77 and 0.31microM for ERalpha and 1.5 and 0.69 microM for ERbeta. These findings indicate that hydroxyanthraquinones such as emodin are phytoestrogens with an affinity to human estrogen receptors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anthraflavic acid, technical grade, 90%