Merck
Infrared matrix-assisted laser desorption/ionization of polycyclic aromatic hydrocarbons with a sulfolane matrix.
Rapid communications in mass spectrometry : RCM (2001-08-17)
S N Jackson, K K Murray
ABSTRAKT
Infrared matrix-assisted laser desorption/ionization (IR-MALDI) of the polyaromatic hydrocarbons (PAHs) anthracene, benzo[a]pyrene, and dibenz[a,h]anthracene was performed using a 10.6-microm CO2 laser and a liquid matrix. Sulfolane (tetrahydrothiophene 1,1-dioxide) was found to be an effective matrix for PAH ionization: mass spectra obtained with a sulfolane matrix contain an intense molecular ion peak; interference from PAH fragment and matrix peaks is negligible in all cases. The main limitation of the sulfolane matrix is sample evaporation after 3 to 5 min in vacuum. This sample lifetime can be increased to between 15 and 30 min using a 2:1 (v/v) mixture of sulfolane and glycerol, but the resulting spectra have greater matrix interference and decreased shot-to-shot signal stability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sulfolane, 99%
Sigma-Aldrich
Dibenz[a,c]anthracene, 97%
Supelco
Sulfolane, analytical standard