Merck
The stability constants of the rare earth complexes with trimetaphosphoric acid.
Annali di chimica (2001-08-18)
S Sungur
ABSTRAKT
The stability constants of the complexes formed between the rare earth metal ions and the anion of trimetaphosphoric acid have been determined at a temperature of 20 degrees C and an ionic strength of mu = 0.1 by the ion-exchange equilibrium method. The investigations indicate that stabilities of complexes increased from La to Lu. The highest and the lowest stability constant values (beta) were found to be 7.76 and 3.82 for Lu3+ and La3+ respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trisodium trimetaphosphate, ≥95%