Merck
The enhancement activities of histidyl-histidine in some prebiotic reactions.
Journal of molecular evolution (1990-01-01)
C Shen, A Lazcano, J Oro
ABSTRAKT
The prebiotic synthesis of His and its dimer has led us to study the possible catalytic properties of His-His. The enhancing effect of His-His has been tested in the dephosphorylation of dAMP, the hydrolysis of oligo(A)12, and the oligomerization of 2'3'-cAMP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adenosine 2′:3′-cyclic monophosphate sodium salt, ≥93%