Merck
The RS-.HSR Hydrogen Bond: acidities of alpha,omega-dithiols and electron affinities of their monoradicals.
Journal of the American Chemical Society (2001-10-05)
J M Karty, Y Wu, J I Brauman
ABSTRAKT
Gas-phase acidities (deltaGo(acid)) have been measured for 1,2-ethanedithiol, 1,3-propanedithiol, and 1,4-butanedithiol, using a Fourier transform ion cyclotron resonance (FT-ICR) mass spectrometer. Adiabatic electron affinities (EAs) of the thiolate monoradicals of these compounds were assigned from electron photodetachment spectra of their corresponding thiolate monoanions, acquired using a cw-ICR. The dithiols exhibit enhanced acidities (up to 8.7 kcal/mol in deltaGo(acid)) and greater EAs (up to 6.7 kcal/mol) than analogous monothiol species. These differences are attributed to an intramolecular RS-.HSR hydrogen bond in the thiolate anion. Considerations of the RO-.HOR hydrogen bond in monoanions of alpha,omega-diols and in the [CH(3)O-.HOCH(3)] complex anion suggest that the RS-.HSR hydrogen bond provides up to 9 kcal/mol extra stabilization.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2-Ethanedithiol, technical grade, ≥90%
Sigma-Aldrich
1,2-Ethanedithiol, ≥98.0% (GC)