Merck
Synthesis of some 2-methylthiouracil nucleosides and its 5-halo analogues of 2-acetamido-2-deoxy-D-glucose.
Die Pharmazie (2001-10-31)
A A Abdel-Rahman
ABSTRAKT
This paper describes the regiospecific synthesis of 5-halo-2-methylthiouracil nucleosides by direct glycosylation followed by halogenation by an electrophilic halogen reagent and sodium azide under mild conditions. The compounds were suggested to be effective as antiviral agents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Methyl-2-thiouracil, purum, ≥98.0% S basis (elemental analysis)
Supelco
6-Methyl-2-thiouracil, VETRANAL®, analytical standard