Merck
  • Committee on Pesticides and Disinfectant Formulations. Pesticide formulations: CIPAC studies. Collaborative International Pesticide Analytical Council.
Committee on Pesticides and Disinfectant Formulations. Pesticide formulations: CIPAC studies. Collaborative International Pesticide Analytical Council.
Journal of AOAC International (2002-03-07)
Alan R Hanks
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Etofenprox, PESTANAL®, analytical standard