Merck
  • Determination of selenium sulfide using 1,3-dibromo-5,5-dimethylhydantoin (DBH). Analytical methods of pharmacopeias with DBH in respect to environmental and economical concern Part 11.
Determination of selenium sulfide using 1,3-dibromo-5,5-dimethylhydantoin (DBH). Analytical methods of pharmacopeias with DBH in respect to environmental and economical concern Part 11.
Journal of pharmaceutical and biomedical analysis (2002-04-04)
Manfred Hilp
ABSTRAKT
USP 2000 (The United States Pharmacopeia, Rockville, MD, 24th ed., 2000, pp. 1514-1515) and PH. EUR. 1997 (European Pharmacopoeia, 3rd ed., Council of Europe, Strasbourg, 1997, p. 1459) dissolve selenium sulfide by boiling with fuming nitric acid for about 1 h. After cooling to room temperature and dilution with water nitrous acid and nitrogen oxides are removed with urea also by boiling before the iodometric titration is performed. This method can be importantly simplified and improved, when using 1,3-dibromo-5,5-dimethylhydantoin (DBH) in glacial acetic acid in presence of solid potassium bromide. Selenium sulfide is dissolved during 5-10 min at room temperature. The excess of DBH and bromine can be removed with 5-sulfosalicylic acid also without boiling. The point of the indicator change for the iodometric titration is improved in comparison to the method of the pharmacopeias, because the colloidal solution of selenium mostly coagulates. Also the sulfur content of selenium sulfide can be determined with DBH.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Dibromo-5,5-dimethylhydantoin, 98%
Sigma-Aldrich
Selenium sulfide