Merck
Occurrence of free and esterified lipoxygenase products in leaves of Glechoma hederacea L. and other Labiatae.
European journal of biochemistry (1989-12-08)
H Kühn, R Wiesner, L Alder, T Schewe
ABSTRAKT
Leaves of Glechoma hederacea L. and other Labiatae contain (9S,10E,12Z,15Z)-9-hydroxy-10,12,15-octadecatrienoic acid, (10E,12Z,15Z)-9-oxo-10,12,15-octadecatrienoic acid, (9S,10E,12Z)-9-hydroxy-10,12-octadecadienoic acid and (10E,12Z)-9-oxo-10,12-octadecadienoic acid in a ratio of 71/14/12/3 (by mass), predominantly esterified in the membrane ester lipids. The leaves contain the highest level of these products, whereas only small amounts were found in the stalk and the roots. The chemical structures of these compounds were established by ultraviolet and infrared spectroscopy, by co-chromatography with authentic standards on various types of HPLC columns including chiral-phase HPLC and gas chromatography/mass spectrometry. The stereochemical specificity indicates the enzymatic origin of the products, most probably via a lipoxygenase reaction. Freshly harvested specimens of G. hederacea L. contain only small amounts of hydroxy-polyenoic fatty acids. Air-drying causes a strong increase in the content of free and esterified (9S,10E,12Z,15Z)-9-hydroxy-10,12,15-octadecatrienoic acid. Up to 80% of the hydroxy fatty acids of the total lipid extracts were esterified in the cellular lipids. The data presented indicate that lipoxygenase products occur in the cellular ester lipids of G. hederacea L. and other Labiatae. The results are discussed in the light of a possible involvement of the lipoxygenase pathway in the natural senescence of leaves.