Merck
Mutations in erg4 affect the sensitivity of Saccharomyces cerevisiae to medium-chain fatty acids.
Biochimica et biophysica acta (2002-05-22)
Virginia McDonough, Joseph Stukey, Trudy Cavanagh
ABSTRAKT
We have found that the medium-chain fatty acids (MCFAs) undecanoic acid (11:0), 10-undecenoic acid (11:1 Delta 10), and lauric acid (12:0) can affect the growth of Saccharomyces cerevisiae in a dose-dependent manner. The principal effect was a longer lag phase in MCFA-containing medium, although higher concentrations of 11:1 Delta 10 inhibited growth. Their relative order of inhibitory action was 11:1 Delta 10>11:0>12:0. Cellular content with MCFA supplementation was dependent on the concentration and the particular species of fatty acid, with 12:0 showing the highest relative accumulation and 11:1 Delta 10 the lowest at all concentrations. We have isolated and characterized a mutant that is hypersensitive to MCFA supplementation and is unable to grow at the normally permissive condition of 1 mM 11:1 Delta 10. However, it does not appear to accumulate higher relative levels of the fed MCFA compared to wild-type cells. Complementation of the mutant revealed that the ERG4 gene, encoding the enzyme that catalyzes the last step in ergosterol biosynthesis, had been mutated. The fatty acid composition of the erg4 Delta mutant differs only slightly from wild-type cells, mainly involving an increase in the relative amount of 12:0. These results indicate that yeast require ergosterol for optimal growth on certain MCFAs. We discuss the role ergosterol may have in cells responding to exogenous MCFAs and in supporting optimal cell growth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Undecanoic acid, ≥99%, FG
Supelco
Undecanoic acid, analytical standard
Sigma-Aldrich
Undecanoic acid, 98%