Merck
Inhibitors of antibiotic efflux pump in resistant Enterobacter aerogenes strains.
Biochemical and biophysical research communications (2002-06-11)
Monique Malléa, Jacqueline Chevalier, Annie Eyraud, Jean-Marie Pagès
ABSTRAKT
Enterobacter aerogenes, a nosocomial pathogen, is frequently exhibiting multidrug resistance mechanisms associated with a change in membrane permeability. In clinical isolates, active efflux plays a prominent role in antibiotic resistance. We report here the effect of three unrelated compounds that are able to restore a noticeable antibiotic susceptibility to resistant strains. The targeting of various parameters which contribute to the efficacy of the efflux mechanism, such as energy, flux selectivity, or functional assembly of the membrane complex, increases the intracellular chloramphenicol concentration in resistant isolates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Globomycin from Streptomyces hagronensis