Merck
Dissipation of the defoliant tribufos in cotton-producing soils.
Journal of agricultural and food chemistry (2002-06-13)
Thomas L Potter, Krishna N Reddy, Eddie P Millhollen, Craig W Bednarz, David D Bosch, Clint C Truman, Timothy Strickland
ABSTRAKT
Soil dissipation of the cotton defoliant tribufos was measured in laboratory incubations and on 0.2-ha research plots. Computed 50% dissipation time (DT(50)) using nonlinear and linear kinetic models ranged from 1 to 19 days. Data indicated that exchangeable soil aluminum inhibited tribufos-degrading soil organisms. Nevertheless, measured DT(50) values were 40 to 700 times less than the aerobic soil half-life (t(1/2)) values used in recent tribufos risk assessments. DT(50) values suggest that risk estimates were overstated. However, edge-of-field runoff concentrations measured on research plots exceeded invertebrate LOECs, thus some aquatic risk is indicated. Field data also suggested that volatilization may be a significant soil dissipation pathway. From this result, we conclude that volatilization should be included in simulation models used for pesticide registration. This will likely improve the accuracy of model outputs for products such as tribufos. Potential volatilization losses indicate a need to evaluate the atmospheric behavior of tribufos.