Merck
Remarkable co-catalysis by copper(I) oxide in the palladium catalyzed cross-coupling of arylboronic acids with ethyl bromoacetate.
Chemical communications (Cambridge, England) (2002-07-18)
Xing-xin Liu, Min-zhi Deng
ABSTRAKT
Copper(I) oxide can effectively co-catalyze the Suzuki type cross-coupling reactions of arylboronic acids with ethyl bromoacetate. As an alternative protocol for introducing the methylenecarboxy group into functionalized molecules, this reaction occurs in the absence of highly toxic thallium compounds or special ligands and should be convenient and practical.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl bromoacetate, reagent grade, 98%
Sigma-Aldrich
Ethyl bromoacetate, purum, ≥97.0% (GC)