Merck
Delayed effects of 1,2-epoxypropyltrimethylammonium chloride on behavioral reactions in rats.
Bulletin of experimental biology and medicine (2002-08-10)
Yu G Plyashkevich, G G Barsegyan
ABSTRAKT
We studied immediate and delayed effects of intraventricular injection of 1,2-epoxypropyltrimethylammonium chloride on behavioral reactions in rats. Apomorphine-induced yawning increased and orientation and exploratory activity was improved 144 h postinjection, which indicates activation of the brain dopaminergic system during this period.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycidyltrimethylammonium chloride, technical, ≥90% (calc. based on dry substance, AT)