Merck
The effect of deuterium and fluorine substitution upon the mutagenicity of N-hydroxy-2,6-dimethylaniline.
Mutation research (2002-09-28)
M Matilde Marques, Gonçalo Gamboa da Costa, Lonnie R Blankenship, Sandra J Culp, Frederick A Beland
ABSTRAKT
2,6-Dimethylaniline (2,6-DMA) is an intermediate in the manufacture of several products, including pesticides, dyestuffs, and synthetic resins. It is also present in nanogram amounts in tobacco smoke, and is a major metabolite of the potent anesthetic and antiarrhythmic drug lidocaine, as well as a nasal carcinogen in rats. As with other aromatic amines, 2,6-DMA can undergo metabolic activation through cytochrome p450-mediated N-hydroxylation, followed by O-esterification to a reactive derivative capable of forming DNA adducts. We have recently characterized four DNA adducts resulting from this metabolic pathway. Three of the adducts arose from reaction of the exocyclic heteroatoms of deoxyadenosine and deoxyguanosine with the carbon para to the arylamine nitrogen. The fourth adduct resulted from reaction of the 2,6-DMA nitrogen with the C8 atom of deoxyguanosine. In order to investigate the relative contribution of the exocyclic heteroatom adducts as compared to the C8-deoxyguanosine adduct to the toxicities elicited by 2,6-DMA, we synthesized and compared the mutagenicity of N-hydroxy-2,6-DMA, N-hydroxy-4-deutero-2,6-DMA, 2,6-dimethylnitrosobenzene, 4-deutero-2,6-dimethylnitrosobenzene, and N-hydroxy-4-fluoro-2,6-DMA. In Salmonella typhimurium TA100, the two deuterated compounds and their non-deuterated analogues gave similar mutagenic responses ( approximately 25 revertants/nmol). Likewise in S. typhimurium TA98, a similar mutant frequency ( approximately 0.7 revertants/nmol) was obtained with the four compounds. With N-hydroxy-4-fluoro-2,6-DMA, the mutant frequency was reduced by approximately 90% in S. typhimurium TA100 and approximately 50% in S. typhimurium TA98. The results suggest that multiple adducts contribute to base substitution mutations detected by S. typhimurium TA100 while the C8-deoxyguanosine adduct is primarily responsible for the frameshift mutations detected by S. typhimurium TA98.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
2,6-Dimethylaniline, analytical standard
Supelco
2,6-Dimethylaniline, PESTANAL®, analytical standard
Sigma-Aldrich
2,6-Dimethylaniline, 99%