Merck
RH-0345 restored partly the effects induced by KK-42 on reproductive events in mealworms.
Mededelingen (Rijksuniversiteit te Gent. Fakulteit van de Landbouwkundige en Toegepaste Biologische Wetenschappen) (2002-11-12)
N Soltani-Mazouni, F Taibi, H Berghiche, G Smagghe, N Soltani
ABSTRAKT
In the mealworm, Tenebrio molitor L., KK-42, an imidazole derivative, was found to reduce ovarian ecdysteroid production, while RH-0345, a benzoylhydrazine analogue, has been reported to affect growth and development of mealworm ovaries in a manner similar to 20-hydroxyecdysone. In order to determine whether the reduction of reproduction events is dependent on inhibition of ecdysteroid production by ovaries, RH-0345 (10 micrograms/insect) was applied before or after topical application of KK-42 (10 micrograms/insect) on newly emerged adult females. KK-42 applied alone increased significantly the pre-oviposition period compared to controls. It also significantly delayed egg-laying compared to combined treatments using KK-42 before or after RH-0345. KK-42 applied alone or with and before RH-0345, was found to reduce significantly the duration of oviposition period. Moreover, there was a significant difference in the duration of oviposition period between the two combined treatments. The fecundity was significantly reduced in all treated series compared to controls. In addition, the fecundity recorded in treated series by RH-0345 alone or before KK-42 was significantly higher than in KK-42 treated series. Lastly, the viability of eggs laid was significantly reduced in all treated series compared to controls. Moreover, the combined treatment pronounced the reduction in egg viability. Free ecdysteroids (ecdysone and 20-hydroxyecdysone) in eggs of control and RH-0345 treated females were analyzed by an enzymo-immunoassay using two specific antibodies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Benzhydrazide, 98%