Merck
Sustaining pattern of phenformin hydrochloride using various polymers and waxes.
Bollettino chimico farmaceutico (2002-12-17)
V P Pandey, N Kannappan, R Manavalan, T Subburaj
ABSTRAKT
The present study was carried out to formulate matrix tablets of phenformin hydrochloride. Granules of phenformin HCl were prepared by using ethyl cellulose, eudragit RS 100, gum acacia, carnauba wax, stearyl alcohol, glyceryl monostearate and triethanol amine. Thus the granules were compressed and fourteen tablets formulations were prepared. All the physical parameters of granules and matrix tablets were studied including compatibility study. One commercial timed disintegration capsule was also included for study and comparison. The results of in vitro studies showed that sustained release matrix tablet might be prepared using carnauba wax, stearyl alcohol, triethanol amine and magnesium stearate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Stearyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Stearyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
1-Octadecanol, Selectophore, ≥99.0%
Sigma-Aldrich
1-Octadecanol, 95%
Sigma-Aldrich
1-Octadecanol, ReagentPlus®, 99%