Merck
Human cytochromes mediating gepirone biotransformation at low substrate concentrations.
Biopharmaceutics & drug disposition (2003-03-06)
David J Greenblatt, Lisa L Von Moltke, Gina M Giancarlo, Daniel A Garteiz
ABSTRAKT
Biotransformation of gepirone to 1-(2-pyrimidinyl)-piperazine (1-PP) and 3'-OH-gepirone, as well as two other hydroxylated metabolites, was studied in vitro using a human liver microsomal preparation and heterologously expressed human CYP3A4 and CYP2D6. The focus was on a low range of gepirone concentrations (1000 nM and below). Liver microsomes formed 1-PP and 3'-OH-gepirone with similar reaction velocities. Two other hydroxylated metabolites (2-OH- and 5-OH-gepirone) were also formed, but pure reference standards were not available for purposes of quantitative analysis. The CYP3A inhibitor ketoconazole completely eliminated 1-PP formation, reduced 3'-OH-gepirone formation to less than 20% of control, and reduced 2-OH-gepirone formation to 7% of control. All metabolites were formed by expressed CYP3A4; however, CYP2D6 formed 3'-OH- and 5-OH-gepirone, but not 1-PP or 2-OH-gepirone. Based on estimated relative abundances of the two isoforms in human liver, CYP3A4 was predicted to account for more than 95% of net clearance of gepirone in vivo at low concentrations approaching the therapeutic range. CYP2D6 would account for less than 5% of net clearance. The findings are consistent with previous in vitro studies of gepirone using higher substrate concentrations.