Merck
[Experience in the use of chemical compounds for the primary identification and differentiation of enteroviruses].
Klinicheskaia laboratornaia diagnostika (2003-04-12)
T V Amvros'eva, V I Votiakov, O T Andreeva, O N Kazinets, Z F Bogush, R M Sharko, Z B Kvacheva
ABSTRAKT
Data on the usage of chemical inhibitors nifan and belvtazid, which possess a selective and antienteroviral effect, in the primary identification of enteroviruses and their differentiation into polio- and non-poliomyelytic ones isolated from human clinical materials or the environment by using the cell culture are presented in the article. The method is recommended for the practical use by the virology service in the diagnostics of enteroviral infections and in the identification of cytopathic agents isolated from the enviroment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Quinolinecarboxylic acid, 97%