Merck
  • The effect of chronic feeding of diacetoxyscirpenol and T-2 toxin on performance, health, small intestinal physiology and antibody production in turkey poults.
The effect of chronic feeding of diacetoxyscirpenol and T-2 toxin on performance, health, small intestinal physiology and antibody production in turkey poults.
British poultry science (2003-05-10)
D Sklan, M Shelly, B Makovsky, A Geyra, E Klipper, A Friedman
ABSTRAKT
1. The effects of feeding T-2 toxin or diacetoxyscirpenol (DAS) at levels up to 1 ppm for 32 d on performance, health, small intestinal physiology and immune response to enteral and parenteral immunisation were examined in young poults. 2. Slight improvement in growth was observed in some groups of poults fed T-2 or DAS mycotoxins for 32 d, with no change in feed efficiency. Feeding both T-2 and DAS resulted in oral lesions which had maximal severity after 7-15 d. 3. Mild intestinal changes were observed at 32 d but no pathological or histopathological lesions were found. Both mycotoxins altered small intestinal morphology, especially in the jejunum where villi were shorter and thinner. In addition, both DAS and T-2 mycotoxins enhanced the proportion of proliferating cells both in the crypts and along the villi. Migration rates were reduced in the jejunum of poults fed T-2 toxin but did not change in the duodenum or in poults fed DAS. 4. No significant effects of T-2 or DAS were observed on antibody production to antigens administered by enteral or parenteral routes. 5. This study indicates that tricothecene toxins at concentrations of up to 1 ppm for more than 30 d influenced small intestinal morphology but did not affect growth or antibody production.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Diacetoxyscirpenol solution, ~100 μg/mL in acetonitrile, analytical standard