- Contact sensitivity to calcium hypochlorite.
Contact sensitivity to calcium hypochlorite.
Contact dermatitis (2003-05-21)
Pankaj S Salphale, Shrutakirthi D Shenoi
PMID12755732
ABSTRAKT
Eusol (an abbreviation for Edinburgh University solution), consisting of aqueous boric acid and calcium hypochlorite, is widely used in the management of open wounds left to heal by secondary intention (1).
MATERIAŁY