Merck
An in vitro gas equilibration method for determination of chemical partition coefficients in fish.
Comparative biochemistry and physiology. A, Comparative physiology (1992-01-01)
A D Hoffman, S L Bertelsen, M L Gargas
ABSTRAKT
1. A gas equilibration method for the determination of in vitro chemical partition coefficients in mammals was adapted for use with fish. 2. In vitro blood: water and tissue: blood partition coefficients were determined for three chlorinated ethanes in rainbow trout (Oncorhynchus mykiss). 3. In vitro partition coefficients accurately predicted chemical concentrations in tissues of exposed trout.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pentachloroethane, 95%