Merck
A denture base resin with low water absorption.
Journal of dentistry (1992-08-01)
M J Barsby
ABSTRAKT
The aim of this study was to investigate poly(isobutyl methacrylate) as a potential denture base material. It was found to have very low water absorption but poor mechanical properties and a low glass transition temperature. The latter features render the material itself unsuitable for use as a denture base material, but indicated that isobutyl methacrylate may be a useful co-monomer for reducing water absorption.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isobutyl methacrylate, 97%