Merck
  • Highly self-organized electron transfer from an iridium complex to p-benzoquinone due to formation of a pi-dimer radical anion complex triply bridged by scandium ions.
Highly self-organized electron transfer from an iridium complex to p-benzoquinone due to formation of a pi-dimer radical anion complex triply bridged by scandium ions.
Journal of the American Chemical Society (2003-10-02)
Junpei Yuasa, Tomoyoshi Suenobu, Shunichi Fukuzumi
ABSTRAKT
Unusually high kinetic order was observed in self-organized Sc3+-promoted electron transfer from tris(2-phenylpyridine)iridium(III) [Ir(ppy)3] to p-benzoquinone (Q) in propionitrile, third-order with respect to the concentration of Sc3+ and second-order with respect to the concentration of Q, to produce a pi-dimer semiquinone radical anion complex that is triply bridged by three Sc3+ ions (Q*--3Sc3+-Q).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propionitrile, 99%
Supelco
Propionitrile, analytical standard
Sigma-Aldrich
2-Phenylpyridine, 98%
Sigma-Aldrich
Propionitrile, purum, ≥99.0% (GC)