Merck
  • Parallelism and dissociation in the actions of an Aroclor 1260-based transformer fluid on testicular androgenesis and antioxidant enzymes.
Parallelism and dissociation in the actions of an Aroclor 1260-based transformer fluid on testicular androgenesis and antioxidant enzymes.
Toxicology (2003-11-26)
Nebojsa L Andric, Silvana A Andric, Sonja N Zoric, Tatjana S Kostic, Stanko S Stojilkovic, Radmila Z Kovacevic
ABSTRAKT
The mechanism by which Aroclors and other polychlorinated biphenyls (PCBs) inhibit testicular androgenesis in vivo and in vitro has not been characterized. Here we studied in adult rats the effects of intratesticular (i.t.t.), intraperitoneal (i.p.) and by gavage (p.o.) administration of Pyralene, an Aroclor 1260-based transformer fluid, on testicular androgenesis and oxidative status in androgen-producing interstitial cells and liver. Pyralene markedly decreased in vitro agonist stimulated androgenesis 24 h after bilateral i.t.t.-injection (25 microg/testis), 24 h and 96 h after single i.p.-injection (10 and 50 mg/kg body weight), and 96 h after p.o.-administration (7x10 mg and 7x50 mg/kg body weight daily). Inhibited androgenesis was accompanied by changes in the activity of antioxidant enzymes (AOEs) in interstitial cells after local i.t.t.-treatment and occasionally after systemic Pyralene application. Among changes in the activity, glutathione peroxidase and catalase reflected relatively well the toxicity of Pyralene in these cells. In liver, glutathione-S-transferase (GST) and glutathione peroxidase activities were enhanced after p.o.-treatment and total glutathione (tGSH) content and lipid peroxidation (LP) were enhanced after i.p.-administration. These results indicate that Pyralene inhibits androgenesis independently of the method of its administration. The results also suggest that changes in the oxidative status in testicular milieu are not critical for Pyralene-induced inhibition of androgenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Aroclor 1260 solution, certified reference material, 500 mg/kg in transformer oil