Merck
[Group intoxication with 2-ethoxyethanol as an ethanol substitute].
Archiwum medycyny sadowej i kryminologii (2003-12-13)
K Reguła, Z Jankowski, K Galer, M Wiergowski, B Szpiech
ABSTRAKT
In this paper the authors presented the group 2-ethoxyethanol intoxication of 8 young men aged between 20 to 22. 50-100 ml of pure compound (diluted with water) was administrated as a drink. In this paper symptoms of acute intoxication to this compound were presented. GC-FID and GC-MS were used as diagnostic tools. The obtained results were compared with the data from available literature.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Ethoxyethanol, ReagentPlus®, 99%
Sigma-Aldrich
2-Ethoxyethanol, spectrophotometric grade, ≥99%
Supelco
2-Ethoxyethanol, analytical standard