Merck
Liposomes composed of reconstituted membranes for induction of tumor-specific immunity.
Methods in enzymology (2004-01-13)
Larry E Westerman, Peter E Jensen
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Octyl β-D-1-thioglucopyranoside, ≥98.0% (GC)