Merck
  • Heteromeric geranyl diphosphate synthase from mint: construction of a functional fusion protein and inhibition by bisphosphonate substrate analogs.
Heteromeric geranyl diphosphate synthase from mint: construction of a functional fusion protein and inhibition by bisphosphonate substrate analogs.
Archives of biochemistry and biophysics (2004-01-17)
Charles Burke, Karin Klettke, Rodney Croteau
ABSTRAKT
Geranyl diphosphate synthase catalyzes the condensation of dimethylallyl diphosphate (C(5)) with isopentenyl diphosphate (C(5)) to produce geranyl diphosphate (C(10)), the essential precursor of monoterpenes. The enzyme from peppermint and spearmint (Menthaxpiperita and Mentha spicata, respectively) functions as a heterodimer or heterotetramer consisting of a 40kDa subunit and 33kDa subunit. The DNAs encoding each subunit were joined with different sized linkers and in both possible orders, and expressed in Escherichia coli to yield the corresponding fused protein. The properties of the recombinant fused version, in which the small subunit was followed by the large subunit with a 10 amino acid linker, resembled those of the native heteromeric enzyme in kinetics, product chain-length specificity, and architecture, and this form thus provided a suitable single gene transcript for biotechnological purposes. Bisphosphonate substrate analogs of the type that inhibit farnesyl diphosphate synthase (C(15)) and geranylgeranyl diphosphate synthase (C(20)) also inhibited the fused geranyl diphosphate synthase, apparently by interacting at both the allylic and homoallylic co-substrate binding sites. The results of inhibition studies, along with the previously established role of the small subunit and related mutagenesis experiments, suggest that geranyl diphosphate synthase employs a different mechanism for chain-length determination than do other short-chain prenyltransferases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Methyl-2-buten-1-ol, 99%
Sigma-Aldrich
3-Methyl-2-buten-1-ol, ≥98%, FG