Merck
Binding site for fungal beta-lactone hymeglusin on cytosolic 3-hydroxy-3-methylglutaryl coenzyme A synthase.
Biochimica et biophysica acta (2004-02-27)
Hiroshi Tomoda, Naomi Ohbayashi, Yuko Morikawa, Hidetoshi Kumagai, Satoshi Omura
ABSTRAKT
We studied the molecular mechanism through which the fungal beta-lactone, hymeglusin, potently and specifically inhibits 3-hydroxy-3-methylglutaryl-CoA (HMG-CoA) synthase. [(14)C]Hymeglusin covalently bound to purified rat liver and to recombinant hamster cytosolic HMG-CoA synthases. The enzyme activity was completely inhibited at a binding ratio of 1.6-2.0 mol [(14)C]hymeglusin/mol HMG-CoA synthase. Incubating the enzyme with 2 mM iodoacetamide (IAA) or 2 mM N-ethylmaleimide (NEM) but not with 1.0 mM diisopropyl fluorophosphates (DFP) completely inhibited the binding, suggesting that hymeglusin binds to a Cys residue of HMG-CoA synthase. Recombinant hamster HMG-CoA synthase labeled with [(3)H]hymeglusin was digested with V8 protease, and the [(3)H]peptide was purified by high performance liquid chromatography (HPLC). The sequence of the peptide was Ser-Gly-Asn-Thr-Asp-Ile-Glu-Gly-Ile-Asp-Thr-Thr-Asn-Ala-[(3)H]hymeglusyl Cys-Tyr-Gly-Gly-Thr-Ala-Ala-Val-Phe-Asn-Ala-Val-Asn-, which corresponds to the active site sequence (from Ser 115 to Asn 141) of hamster HMG-CoA synthase. These findings showed that hymeglusin inhibits hamster cytosolic HMG-CoA synthase by covalently modifying the active Cys 129 residue of the enzyme.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hymeglusin, ≥98% (HPLC), from Fusarium sp.