Merck
Development of a gas chromatographic test for the quantification of the biomarker 3-bromopropionic acid in human urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2004-03-17)
C B'Hymer, K L Cheever
ABSTRAKT
An accurate and precise method was developed for the detection and quantification of 3-bromopropionic acid (3-BPA), a metabolite and biomarker for exposure to 1-bromopropane (1-BP). 1-BP is used as an industrial solvent and exposure is a health concern for industrial workers due to its toxicity. It has been associated with neurological disorders in both animals and humans. Urine sample preparation for the determination of 3-BPA consisted of liquid-liquid extraction (LLE) with ethyl acetate and silylation with N-methyl-N-[tert-butyldimethylsilyl]trifluoroacetamide (MTBSTFA). Quantification was by means of a gas chromatograph (GC) equipped with a mass selective detector (MSD) using a dimethylpolysiloxane (HP-1) capillary column and 3-chloropropionic acid was used as an internal standard in the procedure. Demonstrated accuracy and precision during this method's validation was good; recovery varied between 93 and 98% with relative standard deviations (R.D.S.) of 5.7% or less. The limit of detection (LOD) for the procedure was approximately 0.01microg/ml 3-BPA in urine. These data and other factors of the development and validation of this test method will be discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Bromopropionic acid, 97%