Merck
[Studies on the chemical constituents of Chinese herb Melastoma dodecandrum].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2004-05-14)
Chao Zhang, Yan-xiong Fang
ABSTRAKT
To investigate the chemical constituents of Chinese herb Melastoma dodecandrum. The chemical constituents were isolated by solvent extraction and chromatography. Their structure were identified on the basis of physic-chemical constants and specral data. Five compounds were isolated and identified as hexacosanoic acid (I), quercetin(II), avicularin(III), gallic acid (IV), kaempferol(V). The above-mentioned compounds were separated from the plant for the first time.