Merck
Xanthine oxidase inhibition by 1,3-dipropyl-8-sulfophenylxanthine (DPSPX), an antagonist of adenosine receptors.
Journal of enzyme inhibition and medicinal chemistry (2004-06-19)
Teresa Sousa, Manuela Morato, Eduarda Fernandes, Félix Carvalho, António Albino-Teixeira
ABSTRAKT
Xanthine oxidase (XO), an enzyme involved in purine metabolism, is a source of either oxidants (superoxide radical) or antioxidants (uric acid). Interference with XO activity can lead to oxidative stress, thus contributing to the pathogenesis of cardiovascular diseases. The adenosine receptors antagonist, 1,3-dipropyl-8-sulfophenylxanthine (DPSPX), induces hypertension and cardiovascular injury in rats. Since DPSPX is a xanthine, we aimed at evaluating DPSPX's influence on XO activity to ascertain its contribution to DPSPX-induced hypertension. The activity of isolated XO in the presence of DPSPX was evaluated spectrophotometrically. Serum and urinary uric acid levels of DPSPX-treated rats were measured using a commercial kit. DPSPX inhibited XO activity in a concentration-dependent manner and reduced rat serum and urinary uric acid levels. It can be concluded that: DPSPX is an inhibitor of XO; decreased generation of uric acid may lead to oxidative stress, thus contributing to endothelial dysfunction and vascular morphological changes in DPSPX-treated rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Dipropyl-8-(p-sulfophenyl)xanthine, powder