Merck
  • Degradation of alkyl ethers, aralkyl ethers, and dibenzyl ether by Rhodococcus sp. strain DEE5151, isolated from diethyl ether-containing enrichment cultures.
Degradation of alkyl ethers, aralkyl ethers, and dibenzyl ether by Rhodococcus sp. strain DEE5151, isolated from diethyl ether-containing enrichment cultures.
Applied and environmental microbiology (2004-07-09)
Yong-Hak Kim, Karl-Heinrich Engesser
ABSTRAKT
Twenty strains isolated from sewage sludge were found to degrade various ethers, including alkyl ethers, aralkyl ethers, and dibenzyl ether. In Rhodococcus strain DEE5151, induction of ether degradation needed substrates exhibiting at least one unsubstituted Calpha-methylene moiety as the main structural prerequisite. The cleavage reaction observed with anisole, phenetole, and dibenzyl ether indicates that the initial oxidation occurs at such respective Calpha positions. Diethyl ether-induced strain DEE5151 degraded dibenzyl ether via intermediately accumulated benzoic acid. Phenetole seems to be subject also to another ether-cleaving enzyme. Other strains of this group showed different enzymatic activities towards the substrate classes investigated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dibenzyl ether, purum, ≥98.0% (GC)
Sigma-Aldrich
Benzyl ether, ≥98%, FCC, FG
Sigma-Aldrich
Benzyl ether, 98%